Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Qorpak™ Clear Straight Sided Ultra Clean Round Bottles
Click to view available options
Capacity (Metric):
240 mL
Closure Size:
58-400 mm
Description
- Provide maximum storage capacity and content visibility
- Designed without shoulders
- Made of clear glass
- Cleaned using three-step process to remove traces of bottle release reagents
Order Info
Minimum order quantity is 17 cases.
Specifications
Specifications
Capacity | 8 oz., 240 mL |
Capacity (Metric) | 240 mL |
Capacity (English) | 8 oz. |
Color | Clear |
Diameter (English) | 2.2 in. |
Material | Glass |
Closure Color | Green |
Height (Metric) | 137 mm |
Height (English) | 5.39 in. |
Closure Liner | F217, PTFE |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction