Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Graduated Glass Bottle Beakers with Black Phenolic PolyCone Caps

Catalog No. p-4902971
Click to view available options
Capacity (Metric):
30 mL
60 mL
Closure Size:
33-400 mm
38-400 mm
Packaging:
Bulk Pack
Conventional Pack
Quantity:
288/Cs.
432/Cs.
48/Cs.

For mixing, measuring and storage

  • Designed with slight shoulder for easy pouring and handling
  • Molded graduations in mL and ounces
  • Made of clear glass
  • For use with biological and pathological specimens
  • For storage of industrial laboratory chemicals and reagents

Specifications

Capacity 1 oz., 30mL
Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Clear
Diameter (English) 1.34 in.
Material Glass
Packaging Bulk Pack
Quantity 432/Cs.
Closure Color Black
Height (Metric) 68 mm
Height (English) 2.68 in.
Closure Liner Polycone
Closure Material Phenolic
Closure Size 33-400 mm
Diameter (Metric) 33 mm
Closure Type Screw Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.