Learn More
Qorpak™ Unlined Round Cans with Triple Tite Lid
Permeation protection against light, oxygen, other gases and essential oils
Supplier:  Qorpak™ MET03099
 View more versions of this product
Catalog No. 02-991-239
Add to Cart
Description
Description
  • Features resealable multi-friction plug lid for more than one point of contact between lid and can
  • Provides an airtight seal when opened by prying and sealed by pressing
  • May be sterilized by heat
  • Used for paints, varnishes, pastes and adhesives
Specifications
Specifications
Open Top Can
3785 mL
For paints, varnishes, pastes and adhesives, protection against light, oxygen, other gases and essential oil
Triple Tite Lid, Ears, Bails and Securing Clips
7.5 in.
6.625 in.
128 oz.
Silver
Metal
6 Cs.
708 mm
Product Suggestions
Product Suggestions