Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ White Metal Pulp/Aluminum Foil Lined Caps

Catalog No. 02993366
Click to view available options
Neck Finish:
24-400
33-400
38-400
43-400
48-400
58-400
63-400
70-400
89-400
No. per Case:
144 Cs.
288 Cs.
3000 Cs.
3400 Cs.
576 Cs.
800 Cs.
Packaging:
Bagged
Bulk

Free of odor and taste problems

  • Made of steel coated with anti-corrosive tinplate
  • Impact resistant
  • Tolerate wide ranges of temperature
  • Feature standard 400 GPI thread finish
  • Used for non-acid, non-alkaline products, organic solvents, chrome cleaners, food bottles and jars, brake fluids and mineral oils

Specifications

Color White
Material Metal
No. per Case 576 Cs.
No. per Pack 12/Pk.
Packaging Bagged
Liner Material Pulp/Aluminum Foil
Neck Finish 24-400
For Use With (Equipment) Wide mouth bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.