Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Unlined Polypropylene Caps

Catalog No. 02991122
Click to view available options
Neck Finish:
120-400
33-400
33-430
38-400
43-400
48-400
53-400
58-400
63-400
70-400
89-400
No. per Case:
1100 Caps/Cs.
1300 Caps/Cs.
144 Caps/Cs.
1700 Caps/Cs.
1800 Caps/Cs.
2200 Caps/Cs.
242 Caps/Cs.
2900 Caps/Cs.
4000 Caps/Cs.
456 Caps/Cs.
576 Caps/Cs.
760 Caps/Cs.
900 Caps/Cs.

Polypropylene caps are known for good impact strength, durability, cost-effectiveness, thermal resistance, and pliability.

  • Autoclavable
  • Well-suited for many dry products
  • Add a PTFE disc liner to the unlined polypropylene cap for better leak resistance when autoclaving

Specifications

Color White
Material Polypropylene
No. per Case 456 Caps/Cs.
Packaging Bulk
Neck Finish 89-400
Product Type Ribbed Cap
For Use With (Equipment) Wide Mouth Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.