Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QPCT Monoclonal Antibody (C2), Invitrogen™
GREENER_CHOICE

Catalog No. FEMA551073
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551073 200 μL
FEMA551074 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA551073 Supplier Invitrogen™ Supplier No. MA551073
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GPIP137 regulates the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation/migration in multiple cell types. It binds directly and selectively to MYC and CCND2 RNAs. In neuronal cells, GPIP137 binds to several mRNAs associated with RNA granules including BDNF, CAMK2A, CREB1, MAP2, NTRK2, mRNAs, GRIN1, and KPNB1 mRNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen QPCT
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone C2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene Qpct
Gene Accession No. Q16769
Gene Alias 5730422A13Rik; EC; GCT; Glutaminyl cyclase; glutaminyl-peptide cyclotransferase; glutaminyl-peptide cyclotransferase (glutaminyl cyclase); glutaminyl-tRNA cyclotransferase; Glutamyl cyclase; QC; Qpct; RGD1562284; similar to Glutaminyl-peptide cyclotransferase precursor (QC); sQC
Gene Symbols Qpct
Host Species Mouse
Immunogen Recombinant Human QPCT protein, Val29-Leu361 (Accession #Q16769).
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25797
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.