Learn More
Invitrogen™ QPCT Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593165
Description
Immunogen sequence: MAGGRHRRVV GTLHLLLLVA ALPWASRGVS PSASAWPEEK NYHQPAILNS SALRQIAEGT SISEMWQNDL QPLLIERYPG SPGSYAARQH IMQRIQRLQA DWVLEIDTFL SQTPYGYRSF SNIISTLNPT AKRHLVLACH YDSKYFSHWN NRVFVGATDS AVPCAMMLEL ARALDKKLLS LKTVSDSKPD LSLQLIFFDG EEAFLHWSPQ DSLYGSRHLA AKMASTPHPP GARGTSQLHG MDLLVLLDLI GAPNPTFPNF FPNSARWFER LQAIEHELHE LGLLKDHSLE GRYFQNYSYG GVIQDDHIPF LRRGVPVLHL IPSPFPEVWH TMDDNEENLD ESTIDNLNKI LQVFVLEYLH L; Positive Samples: SW480, Mouse brain, Rat brain.
GPIP137 regulates the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation/migration in multiple cell types. It binds directly and selectively to MYC and CCND2 RNAs. In neuronal cells, GPIP137 binds to several mRNAs associated with RNA granules including BDNF, CAMK2A, CREB1, MAP2, NTRK2, mRNAs, GRIN1, and KPNB1 mRNAs.
Specifications
QPCT | |
Polyclonal | |
Unconjugated | |
Qpct | |
5730422A13Rik; EC; GCT; Glutaminyl cyclase; glutaminyl-peptide cyclotransferase; glutaminyl-peptide cyclotransferase (glutaminyl cyclase); glutaminyl-tRNA cyclotransferase; Glutamyl cyclase; QC; Qpct; RGD1562284; similar to Glutaminyl-peptide cyclotransferase precursor (QC); sQC | |
Rabbit | |
Affinity Chromatography | |
RUO | |
25797, 313837, 70536 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.56 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q16769, Q9CYK2 | |
Qpct | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-361 of human QPCT (NP_0365451). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.