Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QPRT Antibody (24C4), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259433AF405
This item is not returnable.
View return policy
Description
QPRT Monoclonal antibody specifically detects QPRT in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
QPRT | |
Monoclonal | |
Alexa Fluor 405 | |
EC 2.4.2.19, nicotinate-nucleotide pyrophosphorylase (carboxylating), QAPRTase, QPRTasenicotinate-nucleotide pyrophosphorylase [carboxylating], quinolinate phosphoribosyltransferase, Quinolinate phosphoribosyltransferase [decarboxylating] | |
Recombinant human QPRT (1-297aa) purified from E. coli | |
0.1 mL | |
Neuroscience | |
23475 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, ELISA, Immunofluorescence | |
24C4 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction