Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QPRT Antibody (OTI1C10), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273761G
Description
QPRT Monoclonal specifically detects QPRT in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
QPRT | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
QPRT | |
Full length recombinant protein of human QPRT (NP_055113) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1C10 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
EC 2.4.2.19, nicotinate-nucleotide pyrophosphorylase (carboxylating), QAPRTase, QPRTasenicotinate-nucleotide pyrophosphorylase [carboxylating], quinolinate phosphoribosyltransferase, Quinolinate phosphoribosyltransferase [decarboxylating] | |
Mouse | |
Affinity Purified | |
Neuroscience | |
23475.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction