Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QPRT Monoclonal Antibody (OTI7A8)
GREENER_CHOICE

Catalog No. PIMA525213
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525213 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525213 Supplier Invitrogen™ Supplier No. MA525213
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

QPRT Monoclonal Antibody for Western Blot, IHC (P), Flow

QPRT gene encodes a key enzyme in catabolism of quinolinate, an intermediate in the tryptophan-nicotinamide adenine dinucleotide pathway. Quinolinate acts as a most potent endogenous exitotoxin to neurons. Elevation of quinolinate levels in the brain has been linked to the pathogenesis of neurodegenerative disorders such as epilepsy, Alzheimer's disease, and Huntington's disease. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen QPRT
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI7A8
Concentration 0.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene QPRT
Gene Accession No. Q15274
Gene Alias 2410027J01Rik; AI647766; epididymis secretory sperm binding protein Li 90n; HEL-S-90n; nicotinate-nucleotide pyrophosphorylase; nicotinate-nucleotide pyrophosphorylase (carboxylating); nicotinate-nucleotide pyrophosphorylase [carboxylating]; QAPRTase; QPRT; QPRTase; quinolinate phosphoribosyltransferase; Quinolinate phosphoribosyltransferase [decarboxylating]
Gene Symbols QPRT
Host Species Mouse
Immunogen Full length human recombinant protein of QPRT produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23475
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.