Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QPRT Mouse anti-Human, Clone: 24C4, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP25943350UL
Description
QPRT Monoclonal specifically detects QPRT in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
QPRT | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000, Flow Cytometry 1:10-1:1040, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
EC 2.4.2.19, nicotinate-nucleotide pyrophosphorylase (carboxylating), QAPRTase, QPRTasenicotinate-nucleotide pyrophosphorylase [carboxylating], quinolinate phosphoribosyltransferase, Quinolinate phosphoribosyltransferase [decarboxylating] | |
Mouse | |
Protein A purified | |
RUO | |
23475 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a κ |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
24C4 | |
Unconjugated | |
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
QPRT | |
Recombinant human QPRT (1-297aa) purified from E. coli | |
50 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction