Learn More
QPRT Mouse anti-Human, Clone: OTI2H4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501526

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
QPRT gene encodes a key enzyme in catabolism of quinolinate, an intermediate in the tryptophan-nicotinamide adenine dinucleotide pathway. Quinolinate acts as a most potent endogenous exitotoxin to neurons. Elevation of quinolinate levels in the brain has been linked to the pathogenesis of neurodegenerative disorders such as epilepsy, Alzheimer′s disease, and Huntington′s disease. Alternative splicing results in multiple transcript variants.Specifications
QPRT | |
Monoclonal | |
Unconjugated | |
QPRT | |
HEL-S-90n, QPRTase | |
Mouse | |
Affinity Chromatography | |
RUO | |
23475 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI2H4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q15274 | |
QPRT | |
Full length human recombit protein of human QPRT produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.