Learn More
Invitrogen™ QRSL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597088
Description
Immunogen sequence: ELSSEVQSLW SKAADLFESE GAKVIEVSLP HTSYSIVCYH VLCTSEVASN MARFDGLQYG HRCDIDVSTE AMYAATRREG FNDVVRGRIL SGNFFLLKEN YENYFVKAQK VRRLIANDFV NAFNSGVDVL LTPTTLSEAV PYLEFIKEDN RTRSAQDDIF TQAVNMAGLP AVSIPVALSN QGLPIGLQFI GRAFCDQQLL TVAKWFEKQV QFPVIQLQEL MDDCSAVLEN EKLASVSLKQ; Positive Samples: HeLa, Mouse kidney; Cellular Location: Mitochondrion.
Allows the formation of correctly charged Gln-tRNA(Gln) through the transamidation of misacylated Glu-tRNA(Gln) in the mitochondria. The reaction takes place in the presence of glutamine and ATP through an activated gamma-phospho-Glu- tRNA(Gln).
Specifications
QRSL1 | |
Polyclonal | |
Unconjugated | |
QRSL1 | |
2700038P16Rik; C80053; GatA; Glu-AdT subunit A; glu-AdT subunit A {ECO:0000255; glutaminyl-tRNA synthase (glutamine-hydrolyzing)-like 1; glutaminyl-tRNA synthase-like protein 1; glutaminyl-tRNA synthase-like protein 1 {ECO:0000255; glutamyl-tRNA(Gln) amidotransferase subunit A homolog; glutamyl-tRNA(Gln) amidotransferase subunit A, mitochondrial; glutamyl-tRNA(Gln) amidotransferase subunit A, mitochondrial {ECO:0000255; glutamyl-tRNA(Gln) amidotransferase, subunit A; HAMAP-Rule:MF_03150}; QRSL1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55278, 76563 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.98 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9CZN8, Q9H0R6 | |
QRSL1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 289-528 of human QRSL1 (NP_0607623). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.