Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QSER1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP119113AF532
Description
QSER1 Polyclonal antibody specifically detects QSER1 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
QSER1 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
FLJ21924, glutamine and serine rich 1, glutamine and serine-rich protein 1 | |
The immunogen recognized by this antibody maps to a region between residue 1407 ot 1457 of human glutamine and serine rich 1 using the numbering given in entry NP_001070254.1 (GeneID 79832). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
79832 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction