Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QSOX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559881
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559881 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559881 Supplier Invitrogen™ Supplier No. PA559881
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CAEETNSAVC RDFNIPGFPT VRFFKAFTKN GSGAVFPVAG ADVQTLRERL IDALESHHDT WPPACPPLEP AKLEEIDGFF ARNNEEYLAL IF Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 78%.

The QSOX1 gene, also known as Quiescin Q6, is a fusion of two ancient genes: thioredoxin and ERV1. Its expression is induced as fibroblasts begin to exit the proliferative cycle and enter quiescence, suggesting that this gene plays an important role in growth regulation. The QSOX1 protein oxidizes sulfhydryl groups to form disulfide bonds in proteins. QSOX1 expression is induced by hypoxia and appears to protect cells against oxidative stress-induced apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen QSOX1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Qsox1
Gene Accession No. O00391
Gene Alias 1300003H02Rik; FAD-dependent sulfhydryl oxidase-2; hQSOX; mSOx; Q6; Qscn6; Qsox; QSOX1; Quiescin Q6; quiescin Q6 sulfhydryl oxidase 1; quiescin sulfhydryl oxidase 1; RP11-502H18.3; rQSOX; rSOx; Skin sulfhydryl oxidase; Sox; Sox2; Sox-2; Sulfhydryl oxidase 1; testis tissue sperm-binding protein Li 62n; thiol oxidase 1; UNQ2520/PRO6013
Gene Symbols Qsox1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human QSOX1. Recombinant protein control fragment (Product #RP-99851).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5768
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.