Learn More
Invitrogen™ QSOX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120881
Description
Positive test controls include: NCI-H460, U-251MG, Mouse heart. The target is usually found in the following locations: Golgi apparatus membrane, Secreted, Single-pass membrane protein, extracellular space. Immunogen sequence: MRRCNSGSGP PPSLLLLLLW LLAVPGANAA PRSALYSPSD PLTLLQADTV RGAVLGSRSA WAVEFFASWC GHCIAFAPTW KALAEDVKAW RPALYLAALD CAEETNSAVC RDFNIPGFPT VRFFKAFTKN GSGAVFPVAG ADVQTLRERL IDALESHHDT WPPACPPLEP AKLEEIDGFF ARNNEEYLAL IFEKGGSYLG REVALDLSQH KGVAVRRVLN TEANVVRKFG VTDFPSCYLL FRNGSVSRVP VLMESRSFYT AYLQRLSGLT REAAQTTVAP TTANKIAPTV WKLADRSKIY MADLESALHY ILRIEVGRFP VLEGQRLVAL.
The QSOX1 gene, also known as Quiescin Q6, is a fusion of two ancient genes: thioredoxin and ERV1. Its expression is induced as fibroblasts begin to exit the proliferative cycle and enter quiescence, suggesting that this gene plays an important role in growth regulation. The QSOX1 protein oxidizes sulfhydryl groups to form disulfide bonds in proteins. QSOX1 expression is induced by hypoxia and appears to protect cells against oxidative stress-induced apoptosis.
Specifications
QSOX1 | |
Polyclonal | |
Unconjugated | |
Qsox1 | |
1300003H02Rik; FAD-dependent sulfhydryl oxidase-2; hQSOX; mSOx; Q6; Qscn6; Qsox; QSOX1; Quiescin Q6; quiescin Q6 sulfhydryl oxidase 1; quiescin sulfhydryl oxidase 1; RP11-502H18.3; rQSOX; rSOx; Skin sulfhydryl oxidase; Sox; Sox2; Sox-2; Sulfhydryl oxidase 1; testis tissue sperm-binding protein Li 62n; thiol oxidase 1; UNQ2520/PRO6013 | |
Rabbit | |
Affinity chromatography | |
RUO | |
104009, 5768 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.671 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
O00391, Q8BND5 | |
Qsox1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-330 of human QSOX1 (NP_001004128.1). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.