Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QSOX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110775
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110775 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110775 Supplier Invitrogen™ Supplier No. PA5110775
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LWKKHNMVNG RLAGHLSEDP RFPKLQWPTP DLCPACHEEI KGLASWDEGH VLTFLKQHYG RDNLLDTYSA DQGDSSEGGT LARGEEEEKR LTPPEVSHGD RDTQSVRPPG ALGPRPALPE SLHHSLDGKL Highest antigen sequence indentity to the following orthologs: Mouse - 66%, Rat - 65%.

QSOX2 is a member of the sulfhydryl oxidase/quiescin-6 (Q6) family (QSOX1) that regulates the sensitization of neuroblastoma cells for IFN-gamma (IFNG)-induced cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen QSOX2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Qsox2
Gene Accession No. Q6ZRP7
Gene Alias BC030934; neuroblastoma-derived sulfhydryl oxidase; QSCN6L1; QSOX2; quiescin Q6 sulfhydryl oxidase 2; quiescin Q6-like 1; quiescin Q6-like protein 1; quiescin sulfhydryl oxidase 2; SOXN; Sulfhydryl oxidase 2; thiol oxidase 2
Gene Symbols Qsox2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human QSOX2. Recombinant protein control fragment (Product #RP-90405).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 169714
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.