Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QTRT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561795
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561795 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561795 Supplier Invitrogen™ Supplier No. PA561795
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TCLEEMTKRD VPGFAIGGLS GGESKSQFWR MVALSTSRLP KDKPRYLMGV GYATDLVVCV ALGCDMFDCV FPTRTARFGS ALVPTGNLQ Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

tRNA-guanine transglycosylase (TGT; EC 2.4.2.29) synthesizes queuosine (Q), which is found in tRNAs that recognize NAU and NAC codons, encoding tyr, asn, asp, and his. Prokaryotic TGT is a single protein of 43 kD. In contrast, mammalian TGT appears to be a heterodimer consisting of a 60-kD subunit (USP14; MIM 607274) and a 43-kD catalytic subunit (QTRT1) (Deshpande and Katze, 2001 [PubMed 11255023]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen QTRT1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene QTRT1
Gene Accession No. Q9BXR0
Gene Alias 2610028E17Rik; FP3235; Guanine insertion enzyme; QTRT1; queuine tRNA-ribosyltransferase; queuine tRNA-ribosyltransferase 1; queuine tRNA-ribosyltransferase 1 (tRNA-guanine transglycosylase); queuine tRNA-ribosyltransferase catalytic subunit 1; queuine tRNA-ribosyltransferase; queuine tRNA-ribosyltransferase catalytic subunit 1; similar to queuine tRNA-ribosyltransferase 1; Tgt; TGT, 43-KD subunit; TGT, catalytic subunit; TGUT; tRNA-guanine transglycosylase; zgc:66378
Gene Symbols QTRT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human QTRT1. Recombinant protein control fragment (Product #RP-99237).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81890
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.