Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QTRTD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23267425UL
Description
QTRTD1 Polyclonal specifically detects QTRTD1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
QTRTD1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
EC 2.4.2.29, FLJ12960, queuine tRNA-ribosyltransferase domain containing 1, Queuine tRNA-ribosyltransferase domain-containing protein 1, queuine tRNA-ribosyltransferase subunit QTRTD1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
QTRTD1 | |
This antibody was developed against a recombinant protein corresponding to amino acids: FIGMPESLLYCSLHDPVSPCPAGYVTNKSVSVWSVAGRVEMTVSKFMAIQKALQPDWFQCLSDGEVSCKEATS | |
25 μL | |
DNA replication Transcription Translation and Splicing | |
79691 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction