Learn More
BINDER™ Qualification Documents IQ/OQ
SDP
Qualification documents - Digital in PDF format, IQ/OQ documents – supporting documents for validation performed by customers, consisting of: IQ/OQ checklists including calibration guide and comprehensive unit documentation.
Supplier:  BINDER™ 70570002
 View more versions of this product
Catalog No. 08-111-916
Description
Description
  • Parameters depending on unit
  • Available as hardcopy inside folder or PDF document
Specifications
Specifications
BINDER Chambers (Series KBF, KBF-S, KMF, MKF, MKFT)
IQ/OQ Documentation (PDF)
IQ/OQ/PQ with completion of documents
Qualification documents - Digital in PDF format, IQ/OQ documents - supporting documents for validation performed by customers, consisting of: IQ/OQ checklists incl. calibration guide and comprehensive unit documentation; parameters: temperature and humidity values
BINDER Chambers
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.