Learn More
BINDER™ Qualification Documents IQ/OQ
Shop All BINDER Inc. Products
Click to view available options
For Use With (Equipment):
BINDER Chambers (Series BD, BF, ED, FD, FED, FDL, FP, M, C, CB, KB, KT, MK, MKT, VD, VDL, UF V)
Description
- Parameters depending on unit
- Available as hardcopy inside folder or PDF document
Specifications
Specifications
| For Use With (Equipment) | BINDER Chambers (Series BD, BF, ED, FD, FED, FDL, FP, M, C, CB, KB, KT, MK, MKT, VD, VDL, UF V) |
| Description | Qualification documents - Digital in PDF format, IQ/OQ documents - supporting documents for validation performed by customers, consisting of: IQ/OQ checklists incl. calibration guide and comprehensive unit documentation; parameters: temperature, CO2, O2, pressure, depending on unit |
| Product Type | IQ/OQ Documentation (PDF) |
| Product Line | BINDER Chambers |
| Service Offering | IQ/OQ/PQ with completion of documents |