Learn More
Description
- For use with liquid clarification, qualitative analytical separations for precipitates, buffers filtration, soil analysis, food and beverage testing
- High purity filters
- Consistent performance
- Available in a variety of sizes
Specifications
Specifications
| Filter Type | Ashless |
| Format | Disc |
| Material | 100% Pure Cotton Linter. Ashless (0.007%) |
| Filtration Speed | Medium Speed |
| Flow Rate | Medium |
| For Use With (Application) | Ideal for gravimetric analysis of components found in industries like mining or steel. Ideal as a high purity filter for trace element analysis. |
| Grade | Grade 74 |
| Quantity | 100 Pk. |
| Retention | 2 um |
| Type | Qualitative Filter Paper |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.