Learn More
Ahlstrom-Munksjo Qualitative Filter Paper
Catalog No. 12007235 Shop All Ahlstrom Products
Change view
Click to view available options
Filter Type:
Ashless
Hardened Ashless
Low Ash
Diameter (Metric):
21 mm
47 mm
55 mm
70 mm
90 mm
110 mm
125 mm
185 mm
240 mm
270 mm
320 mm
400 mm
425 mm
500 mm
Filtration Speed:
Fast Speed
Medium Speed
Slow Speed
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filter Type Diameter (Metric) Filtration Speed
12-007-235 Ashless 320 mm Medium Speed
12-007-196 Low Ash 47 mm Fast Speed
12-007-198 Low Ash 70 mm Fast Speed
12-007-200 Low Ash 110 mm Fast Speed
12-007-201 Low Ash 125 mm Fast Speed
12-007-203 Low Ash 185 mm Fast Speed
12-007-207 Low Ash 400 mm Fast Speed
12-007-220 Hardened Ashless 240 mm Fast Speed
12-007-226 Ashless 55 mm Medium Speed
12-007-228 Ashless 90 mm Medium Speed
12-007-237 Hardened Ashless 21 mm Medium Speed
12-007-239 Hardened Ashless 425 mm Medium Speed
12-007-247 Hardened Ashless 240 mm Medium Speed
12-007-249 Hardened Ashless 320 mm Medium Speed
12-007-251 Hardened Ashless 500 mm Medium Speed
12-007-266 Hardened Ashless 425 mm Slow Speed
12-007-273 Hardened Ashless 185 mm Slow Speed
12-007-275 Hardened Ashless 270 mm Slow Speed
12-007-295 Low Ash 70 mm Slow Speed
12-007-297 Low Ash 110 mm Slow Speed
12-007-309 Low Ash 90 mm Medium Speed
12-007-311 Low Ash 125 mm Medium Speed
12-007-313 Low Ash 185 mm Medium Speed
12-007-315 Low Ash 270 mm Medium Speed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
Catalog No. 12-007-235 Supplier Ahlstrom-Munksjo Supplier No. 07403200
Add to Cart
Add to Cart
Ahlstrom-Munksjo qualitative filter paper is recommended for use in analytical methods, which determine or identify particulate constituents of a mixture irrespective of the amount present. They are often used in routine separation work that still requires high purity and consistent performance.
  • For use with liquid clarification, qualitative analytical separations for precipitates, buffers filtration, soil analysis, food and beverage testing
  • High purity filters
  • Consistent performance
  • Available in a variety of sizes
TRUSTED_SUSTAINABILITY
Specifications
Filter Type Ashless
Format Disc
Filtration Speed Medium Speed
Type Qualitative Filter Paper
Diameter (Metric) 320 mm