Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
- Home
- Products
- Filters and Filtration
- Filter Paper
- Ahlstrom-Munksjo Qualitative Filter Paper
Ahlstrom-Munksjo Qualitative Filter Paper
Catalog No. 12007237 Shop All Ahlstrom Products
Change view
Click to view available options
Filter Type:
Ashless
Hardened Ashless
Low Ash
Diameter (Metric):
21 mm
47 mm
55 mm
70 mm
90 mm
110 mm
125 mm
185 mm
240 mm
270 mm
320 mm
400 mm
425 mm
500 mm
Filtration Speed:
Fast Speed
Medium Speed
Slow Speed
24 product options available for selection
| Catalog No. | Filter Type | Diameter (Metric) | Filtration Speed |
|---|---|---|---|
| 12-007-237 | Hardened Ashless | 21 mm | Medium Speed |
| 12-007-196 | Low Ash | 47 mm | Fast Speed |
| 12-007-198 | Low Ash | 70 mm | Fast Speed |
| 12-007-200 | Low Ash | 110 mm | Fast Speed |
| 12-007-201 | Low Ash | 125 mm | Fast Speed |
| 12-007-203 | Low Ash | 185 mm | Fast Speed |
| 12-007-207 | Low Ash | 400 mm | Fast Speed |
| 12-007-220 | Hardened Ashless | 240 mm | Fast Speed |
| 12-007-226 | Ashless | 55 mm | Medium Speed |
| 12-007-228 | Ashless | 90 mm | Medium Speed |
| 12-007-235 | Ashless | 320 mm | Medium Speed |
| 12-007-239 | Hardened Ashless | 425 mm | Medium Speed |
| 12-007-247 | Hardened Ashless | 240 mm | Medium Speed |
| 12-007-249 | Hardened Ashless | 320 mm | Medium Speed |
| 12-007-251 | Hardened Ashless | 500 mm | Medium Speed |
| 12-007-266 | Hardened Ashless | 425 mm | Slow Speed |
| 12-007-273 | Hardened Ashless | 185 mm | Slow Speed |
| 12-007-275 | Hardened Ashless | 270 mm | Slow Speed |
| 12-007-295 | Low Ash | 70 mm | Slow Speed |
| 12-007-297 | Low Ash | 110 mm | Slow Speed |
| 12-007-309 | Low Ash | 90 mm | Medium Speed |
| 12-007-311 | Low Ash | 125 mm | Medium Speed |
| 12-007-313 | Low Ash | 185 mm | Medium Speed |
| 12-007-315 | Low Ash | 270 mm | Medium Speed |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
Description
- For use with liquid clarification, qualitative analytical separations for precipitates, buffers filtration, soil analysis, food and beverage testing
- High purity filters
- Consistent performance
- Available in a variety of sizes
Specifications
Specifications
| Filter Type | Hardened Ashless |
| Format | Disc |
| Filtration Speed | Medium Speed |
| Type | Qualitative Filter Paper |
| Diameter (Metric) | 21 mm |
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?