Learn More
Invitrogen™ Quant-iT™ 1X dsDNA Assay Kits, high sensitivity (HS) and broad range (BR)

Description
Make DNA quantitation easy and accurate in a ready-to-go 1X formulation with the Quant-iT 1X dsDNA HS (High-Sensitivity) and Quant-iT 1X dsDNA BR (Broad Range) Assay kits. Both kits provide a ready-to-use working solution. Simply load your sample, add the Qubit 1X dsDNA HS or BR working solution, and read the concentration using a fluorescence-based microplate reader.
The Quant-iT 1X dsDNA HS Assay Kit is accurate for initial sample concentrations ranging from 10 pg/μL to 100 ng/μL, providing a core detection range of 0.2 ng to 100 ng of DNA in the assay tube. The Quant-iT 1X dsDNA BR Assay Kit is accurate for initial DNA sample concentrations of 200 pg/μL to 4000 ng/μL (depending on sample volume), providing a detection range of 4–4000 ng of DNA.
Common contaminants such as salts, free nucleotides, solvents, detergents, and protein are well tolerated in the dsDNA assays. The '1X' dsDNA assay kits provide reagent and buffer in a ready-to-use formulation and are highly selective for dsDNA over ssDNA, RNA, and protein.
Specifications
Specifications
| Assay | 1 x dsDNA, high sensitivity |
| Detection Method | Fluorescence |
| Format | Kit |
| Sample Type | DNA |
| For Use With (Equipment) | Microplate Reader |
| Quantitation Range | 0.2 to 100 ng |
| Product Line | Quant-iT |
| Quantity | 1000 assays |
| Shipping Condition | Wet Ice |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.