Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Quant-iT™ OliGreen™ ssDNA Assay Kit and Quant-iT OliGreen ssDNA Reagent

Catalog No. o11492
Encompass_Preferred
Click to view available options
Quantity:
1 kit
1 mL
Product Type:
Quant-iT OliGreen ssDNA Kit
Quant-iT OliGreen ssDNA Rgnt

Includes

Kit contains an ultrasensitive, green-fluorescent nucleic acid stain, buffer and an oligonucleotide standard. OliGreen reagent also available separately (Cat. No. O7582).

Quant-iT™ OliGreen ssDNA Assay Kit and Quant-iT OliGreen ssDNA Reagent contain an ultrasensitive, green-fluorescent nucleic acid stain for quantitating oligonucleotides and single-stranded DNA (ssDNA) in solution.

Quantify ssDNA with ease using the Quant-iT OliGreen ssDNA Assay kit and reagent. This assay kit and reagent enable sensitive quantitation of single-stranded DNA without the need for a highly concentrated sample. As little as 100 pg/mL can be quantitated using the assay kit or Quant-iT OliGreen ssDNA Reagent.

Quantitate ssDNA and other oligonucleotides accurately and without the need for a highly concentrated sample with Quant-iT OliGreen ssDNA Assay Kit and Quant-iT OliGreen ssDNA Reagent. Quant-iT OliGreen ssDNA Assay Kits contain an ultrasensitive, green-fluorescent nucleic acid stain for quantitating oligonucleotides and single-stranded DNA (ssDNA) in solution. The Quant-iT™ OliGreen™ ssDNA Assay Reagent enables researchers to quantify as little as 1 ng/mL oligonucleotide or ssDNA (200 pg in a 200 μL assay volume) with a fluorescence microplate reader using fluorescein excitation and emission wavelengths. This sensitivity exceeds that achieved with absorbance methods by 10,000-fold.

In addition to the dye, the OliGreen ssDNA Assay Kit includes buffer and an oligonucleotide standard. The OliGreen reagent is also available as the stand-alone Quant-iT OliGreen ssDNA Reagent.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Detection Method Fluorescence
Excitation/Emission 500/525
For Use With (Equipment) Microplate Reader
No. of Reactions 200 assays (2 mL assay volume)
Quantitation Range 200 pg to 2 μg
Product Line OLIGREEN, Quant-iT
Product Type Quant-iT OliGreen ssDNA Kit
Quantity 1 kit
Shipping Condition Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.