Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Quanti-Cult Plus™ Aspergillus brasiliensis ATCC™ 16404™

Catalog No. r4711100
Click to view available options
Quantity:
100 Tests/Kit
This item is not returnable. View return policy

Catalog No. R4711100

Supplier: Thermo Scientific™ R4711100

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

10 vials (100 tests)

Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.

Thermo Scientific™ Quanti-Cult Plus™ Aspergillus brasiliensis ATCC™ 16404™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial.

  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.1mL inoculum
  • 10 tests/reconstituted vial; 100 tests/kit
  • Ready for rehydration
  • For use in quality control procedures

Includes:

10 preserved organism vials and 10 rehydration vials.

TRUSTED_SUSTAINABILITY

Specifications

ATCC® 16404TM*
Description Aspergillus brasiliensis ATCC™ 16404™
Format Quanti-Cult Plus
Product Type Quality Control Organism
Target Organism Class Fungi
Test Type Quantitative QC
Packaging Type 100 tests (10 vials x 10 tests ea.)
Quantity 100 Tests/Kit
Strain Designations WLRI 034(120) [CBS 733.88, DSM 1387, DSM 1988, IFO 9455, IMI 149007, NCPF 2275]

*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.