Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Quanti-Cult™ Aspergillus brasiliensis ATCC™ 16404™

Catalog No. r4731100
Click to view available options
Description:
Aspergillus brasiliensis ATCC™ 16404™
This item is not returnable. View return policy

Catalog No. R4731100

Supplier: Thermo Scientific™ R4731100

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.

Thermo Scientific™ Quanti-Cult™ Aspergillus brasiliensis ATCC™ 16404™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial, ready for rehydration and use in quality control procedures.

  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.3mL inoculum

Ordering Alerts:

1 Test/reconstituted vial; 10 Tests/kit

Includes:

10 preserved organism vials and 10 rehydration vials.

TRUSTED_SUSTAINABILITY

Specifications

ATCC® 16404TM*
Description Aspergillus brasiliensis ATCC™ 16404™
Format Quanti-Cult
Product Type Bactidisk
Target Organism Class Fungi
Test Type Quantitative QC
Packaging Type 10 Test/Kit
Strain Designations WLRI 034(120) [CBS 733.88, DSM 1387, DSM 1988, IFO 9455, IMI 149007, NCPF 2275]

*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.