Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Quanti-Cult Plus™ Burkholderia cenocepacia ATCC™ BAA-245™

Catalog No. R4715221
Change view
Click to view available options
Quantity:
100 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R4715221 100 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R4715221 Supplier Thermo Scientific™ Supplier No. R4715221
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy

Quality control to comply with USP chapter 60

Thermo Scientific™ Quanti-Cult Plus™ Burkholderia cenocepacia ATCC™ BAA-245™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures. It is the recommended strain to support QC testing for non-sterile products described in US Pharmacopeia chapter 60.

  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.1mL inoculum
  • 10 tests/reconstituted vial; 100 tests/kit
  • Ready for rehydration
  • For use in quality control procedures
Includes: 10 preserved organism vials and 10 rehydration vials.
TRUSTED_SUSTAINABILITY

Specifications

ATCC® ATCC™ BAA-245™
Description Burkholderia cenocepacia ATCC™ BAA-245™
Format Quanti-Cult Plus™
Product Type Bactidisk
Target Organism Class Gram-negative Rods
Test Type Quantitative QC
For Use With (Application) Pharmaceutical
Packaging Type 100 Test/Kit
Quantity 100 Tests/Kit
Strain Designations LMG 16656

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.