Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Quanti-Cult Plus™ Staphylococcus aureus subsp. aureus ATCC™ 6538P™

Catalog No. r4719002
Click to view available options
Quantity:
100 Tests/Kit
This item is not returnable. View return policy
This item is not returnable. View return policy

Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.

Thermo Scientific™ Quanti-Cult Plus™ Staphylococcus aureus subsp. aureus ATCC™ 6538P™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.

  • No serial dilution required
  • Delivers less-than100 colony forming units (CFUs) in a single 0.1mL inoculum
  • 10 tests/reconstituted vial; 100 tests/kit
  • Ready for rehydration
  • For use in quality control procedures
According to ATCC S. aureus ATCC™ 6538P™ was found moderately resistant to penicillin in 1944 using a Penicillin P2 disk. This result was confirmed in initial characterization at ATCC, but subsequent investigation using a Penicillin P10 disk did not show resistance. Resistance has not been confirmed using modern methods in accordance with the Clinical & Laboratory Standards Institute guidelines.

Includes:

10 preserved organism vials and 10 rehydration vials.

 

TRUSTED_SUSTAINABILITY

Specifications

ATCC® 6538PTM*
Description Staphylococcus aureus subsp. aureus ATCC™ 6538P™
Format Quanti-Cult Plus
Product Type Quality Control Organism
Target Organism Class Cocci
Test Type Quantitative QC
Packaging Type 100 Test/Kit
Quantity 100 Tests/Kit
Strain Designations FDA 209P

*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.