Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega QuantiFluor™ ONE dsDNA System

Catalog No. pre4870
Click to view available options
No. of Reactions:
100
500

Includes

  • QuantiFluor ONE dsDNA Dye
  • QuantiFluor ONE Lambda DNA
  • 1X TE Buffer, pH 7.5

The QuantiFluor(R) ONE dsDNA System enables sensitive quantitation of small amounts of double-stranded DNA (dsDNA). The system contains a fluorescent dsDNA-binding dye (504nm[Ex]/531nm[Em]) developed for use in an add-and-read format for dye and standard.

  • Fluorescent Quantitation of dsDNA with Add-and-Read Convenience
  • Specific quantitation of dsDNA
  • Increased sensitivity over absorbance at 260nm for low-concentration samples
  • Simple to use and automation-friendly

Specifications

Content And Storage Varies Among Components
No. of Reactions 500
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.