Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuantiGene™ Plex Assay Kit

Catalog No. QP1014
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Plate
10 Plates
3 Plates
Catalog No. Quantity
QP1014 3 Plates
QP1013 1 Plate
QP1015 10 Plates
3 options

Catalog No. QP1014

Supplier: Invitrogen QP1014

Only null left
Add to Cart
Add to Cart

The QuantiGene Plex Assay Kit contains the generic reagents, buffers, and plates necessary to perform the assay in 96-well plates and must be used with a target-specific QuantiGene Plex panel (sold separately).

The QuantiGene Plex Assay is a gene expression assay that enables multiplexed measurement of up to 80 RNA transcript targets. The assay uses target-specific probes to capture the RNA of interest, and branched DNA technology then amplifies fluorescent signals which are read with a Luminex xMAP instrument.

Features of the QuantiGene Plex Assay include:

  • Difficult sample types—works with degraded and cross-linked RNA in FFPE tissues and blood
  • True multiplexing—measure up to 80 genes of interest and housekeeping genes in the same well with no cross-reactivity
  • Standardized platform—uses 96 or 384-well plate formats compatible with the INTELLIFLEX, Luminex 200, FlexMap3D, and MagPix systems
  • Simple workflow—employs an ELISA-like workflow for direct hybridization of transcripts to beads and transcript labeling
  • Large inventory of validated genes—select from over 15,000 genes to create pathway and disease-themed panels
  • Rapid turnaround for custom panels—if we don't have your gene(s), we can create a custom panel within two weeks

Technology

QuantiGene assays utilize branched DNA (bDNA) technology that has been used for decades in the VERSANT™ diagnostic assays for HIV-1, HCV, and HBV. First, cells are lysed or tissue samples are homogenized to release the target RNA. Second, the oligonucleotide probe set is incubated with the target RNA and Luminex xMAP bead set overnight. During this incubation, the probes cooperatively hybridize to the target and capture probes bound to each bead, capturing the target RNA. Third, signal amplification is performed via sequential hybridization of the bDNA preamplifier, amplifier, and label-probe molecules to the target. Addition of streptavidin phycoerythrin generates a luminescent signal directly proportional to the amount of target mRNA present in the sample.

Applications

QuantiGene Plex Assay applications include compound screening of drug candidates, biomarker verification, siRNA knockdown efficiency, prospective and/or retrospective analysis of clinical samples, miRNA profiling, microarray validation, predictive toxicology, and detection of translocations and fusion genes.

TRUSTED_SUSTAINABILITY

Specifications

Format 96-well
For Use With (Application) QuantiGene Plex Panel
For Use With (Equipment) Luminex instrument
Content And Storage

• 105 μL Proteinase K, –20°C
• 810 μL Blocking Reagent, –20°C
• 34.5 mL Label Probe Solution, 2–8°C
• 34.5 mL Pre-Amplifier Solution, 2–8°C
• 34.5 mL Amplifier Solution, 2–8°C
• 34.5 mL SAPE Solution, 2–8°C
• 36 mL Lysis Mixture, 15–30°C
• 2.4 mL Wash Buffer Component 1, 15–30°C
• 39 mL Wash Buffer Component 2, 15–30°C
• 34.5 mL10x SAPE Wash Buffer Concentrate, 15–30°C
• 3 Magnetic Separation Plates, 15–30°C
• 30 Plate Seals, 15–30°C
• 3 Hybridization Plates, 15–30°C
• 6 Pressure Seals, 15–30°C

Note: Prepared 1x Wash Buffers can be stored for 1 week protected from light at 4°C.

Product Line QuantiGene Plex
Product Type Multiplex Assay Kit
Quantity 3 Plates
Shipping Condition Blue Ice
Sample Type Purified RNA, cell/blood/FFPE lysates, tissue homogenates
Detection Method Fluorescence
Species Universal
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.