Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QuantiGene™ Plex Wash Buffer Components

Click to view available options
Quantity:
1 Plate
10 Plates
3 Plates
Description
QuantiGene Plex Wash Buffer Components are aqueous buffered solutions that are mixed with nuclease-free water to make QuantiGene Plex wash buffer. The wash buffer components may be purchased in 1, 3, or 10-plate quantities to supplement QuantiGene Plex Assay kits when additional wash buffer volume is required for automation.

Specifications
Specifications
Format | 96-well |
For Use With (Application) | QuantiGene Plex Assay |
Content And Storage | • 0.8 mL Wash Buffer Component 1 • 13 mL Wash Buffer Component 2 Store at room temperature. |
Key Functions | Multi-Gene Expression |
Product Line | QuantiGene Plex |
Product Type | Wash Buffer |
Quantity | 1 Plate |
Shipping Condition | Room Temperature |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction