Learn More
R&D Systems™ Quantikine Immunoassay Control 1117 for Human IFN-a 2 HS
Shop All Novus Biologicals Products

Description
Quantikine Immunoassay Control 1117 designed for use with Human IFN-α 2 HS.
- Species: Human
- Storage: 2°C to 8°C
- Shipping: Ambient temperature
- Research Area: Interferons
- For Use With: HSIFNA2
Specifications
Specifications
| Product Type | ELISA Control |
| Quantity | 1 Each |
| Species | Human |
| Binding Target | IFN-alpha 2 |
| Gene ID (Entrez) | 3440 |
| Product Line | R&D Systems |
| Synonym | alpha-2a interferon, Ifa2, IFNA, IFNA2, IFNA2a, IFNA2b, IFNA2c, IFNAA, IFNalpha 2, IFN-alpha-2, IFN-alphaA, INFA2, interferon alpha 2b, interferon alpha A, interferon alpha-2, Interferon alpha-A, interferon, alpha 2, LeIF A, MGC125764, MGC125765 |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Research Area | Cytokines |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.