Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Quantikine Immunoassay Control Set 248 for Human TNF-alpha

Catalog No. qc248
Click to view available options
Quantity:
1 Each
This item is not returnable. View return policy
This item is not returnable. View return policy

For use with catalog numbers DTA00D, STA00D, and PDTA00D

For use as quantitative controls for the determination of cytokine concentrations in biological fluids. Concentrations have been assigned using R&D Systems™’ Quantikine™ ELISA kits. Controls are prepared in a buffered protein base with preservatives. They contain recombinant human cytokines at low, medium and high concentrations. Controls are supplied lyophilized.

Specifications

Content And Storage Store the unopened product at 2-8°C. Do not use past expiration date.
Gene ID (Entrez) 7124
Quantity 1 Each
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.