Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Quantikine Immunoassay Control Group 8
Click to view available options
Quantity:
1 Each
Description
- For use as quantitative controls for the determination of cytokine concentrations in biological fluids
- Concentrations have been assigned using R&D Systems’ Quantikine™ kits
- Controls are prepared in diluted porcine serum with preservatives
- They contain recombinant human cytokines at low, medium and high concentrations
- Controls are supplied lyophilized
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Type | Quantikine Immunoassay Control Group 8 |
| Intended Use | Control |
| Quantity | 1 Each |
| Stability | Stable for at least 6 months from date of receipt |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction