Learn More
Description
Quantikine Immunoassay Control Group QC321 contains controls for human epidermal growth factor (EGF). These controls are designed for use with R&D Systems Quantikine ELISA kits.
- Analyte: Human EGF
- Storage: Store unopened at 2°C to 8°C
- Reconstitution: Reconstitute with 3.0 mL of deionized or distilled water
- Stability: Stable for at least 6 months from date of receipt when stored at 2°C to 8°C
- Usage: For research use only
Specifications
Specifications
| Product Type | ELISA Control |
| Quantity | 1 Ea. |
| Species | Human |
| Binding Target | EGF |
| Gene ID (Entrez) | 1950 |
| Product Line | R&D Systems |
| Synonym | beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), hEGF, HOMG4, pro-epidermal growth factor, URG, Urogastrone |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Research Area | Growth Factors |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
