Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quantitative Analysis Spectroscope
SCI_ED

Catalog No. s42581a
Change view
Click to view available options
:
20cmL
Catalog No.
S42581A 20cmL
1 options
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Catalog No. S42581A

Supplier: Envision System Inc 10010

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Includes

Instructions and suggestions for laboratory use

Highly accurate diffraction grating spectroscope.

  • Housed in an unbreakable plastic case
  • Wavelength range: 4000 to 7000Å
  • Accuracy: ±50Å
  • Overall length: 7.88 in. (20cm)
  • Scale length: 1.5 in. (37mm)

Specifications

For Use With (Application) Highly accurate diffraction grating spectroscope
Wavelength 400 to 700 nm
Product Type Quantitative Spectroscope
Includes Instructions and suggestions
Length (Metric) 20 cm
Length (English) 7.88 in.
Material Plastic
Accuracy ±50A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.