Learn More
Quantitative Electroscope
Ideal for student electrostatic experiments.
Supplier: Science First 14435
Description
- Measures conducted and inducted charges
- When the electroscope is charged, the conductively coated plastic straw rotates in proportion to the magnitude of the charge
Specifications
Student electrostatic experiments and is used to make quantitative measurement of electric charge, study the photoelectric effect, and investigate the behavior of conductors and insulators | |
Quantitative Electroscope |