Learn More
Eisco™ Quartz Mineral Specimen
SCI_ED
Catalog No. S26633 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S26633 10/Pk.
S24671 1 Ea.
S24672 12/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S26633 Supplier Eisco™ Supplier No. ESNG0010PK10
Add to Cart
Add to Cart
Quartz mineral specimen for use in classroom identification labs and exercises.
  • Useful during lessons on mineral hardness, luster, and fracture
  • Has hardness of 7, complete lack of cleavage planes, and distinct euhedral crystal habit
Specifications
Product Type Mineral
Quantity 10/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg
Specimen Type Quartz