Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Qubit™ Fluorometer International Power Supply

Catalog No. A36204
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A36204 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A36204 Supplier Invitrogen™ Supplier No. A36204
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Qubit Fluorometer International Power Supply is designed for use with either the Qubit 3 or Qubit 4 Fluorometer.

The Qubit Fluorometer International Power Supply is designed for use with either the Qubit 3 or Qubit 4 Fluorometer. This power supply should not be used with the original or Qubit 2.0 Fluorometer. The product includes a power cord and four interchangeable adaptors and enables the Qubit to be plugged into outlets around the world.

TRUSTED_SUSTAINABILITY

Specifications

Product Line Qubit
Shipping Condition Room Temperature
Type Power Supply
Quantity 1 each
For Use With (Equipment) Qubit 3 Fluorometer, Qubit 4 Fluorometer

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.