Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Qubit™ Fluorometer USB Flash Drive

Catalog No. Q46009
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q46009 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q46009 Supplier Invitrogen™ Supplier No. Q46009
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Qubit Fluorometer USB Flash Drive is designed for the transfer of data from the Qubit Fluorometer.

The Qubit Fluorometer USB Flash Drive is designed for the transfer of data from the Qubit Fluorometer. Only the Qubit 2.0, 3, and 4 fluorometers have an available USB port. Data exported from the Qubit Fluorometer are stored in CSV format and can be opened by programs such as Microsoft Excel.

TRUSTED_SUSTAINABILITY

Specifications

Product Line Qubit
Type USB Flash Drive
Shipping Condition Room Temperature
Quantity 1 each
For Use With (Equipment) Qubit Fluorometer

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.