Learn More
Epredia™ Quick Release Clamp, Aligner
Catalog No. 715020s
Click to view available options
Product Type:
Universal Cassette Clamp
Fix cassettes or embedding rings both longitudinally and horizontally, with orientation on X-, Y-, and Z-axes using the Epredia™ Quick Release Clamp.
Universal Cassette Clamps
  • For all types of commercially available cassettes
  • Silver model is for HM 325, HM 355S, and HM 340E models with serial numbers above 34000
Quick Release Clamp
  • For embedding rings
Zero Orientation Device
  • Positions specimen holder flat against the knife or blade (aligns X, Y axis)
Specifications
Product Type Universal Cassette Clamp
For Use With (Equipment) Cassettes, Embedding Rings
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.