Learn More
R&D Systems™ QuicKit Control for Human/Cynomolgus Monkey PD-L1/B7-H1
Shop All Novus Biologicals Products

Description
QuicKit Control for Human and Cynomolgus Monkey PD-L1/B7-H1 provides quantitative assessment of protein levels in ELISA assays.
- Detection Method: Colorimetric - 450 nm (TMB)
- Species Reactivity: Human, Cynomolgus Monkey
- Measurement: Quantitative ELISA
- Reconstitution: Reconstitute with deionized or distilled water; Low Control requires 1.5 mL, High Control requires 2.5 mL
- Storage: Store unopened product at 2°C to 8°C; stable for at least 6 months from date of receipt
- Shipping: Ambient temperature
Specifications
Specifications
| Product Type | ELISA Control |
| Quantity | 1 Ea. |
| Species | Human, Cynomolgus Monkey |
| Binding Target | PD-L1 |
| Gene ID (Entrez) | 29126 |
| Product Line | R&D Systems |
| Synonym | Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Research Area | Immunology |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.