Learn More
Description
QuicKit Immunoassay Control for Human Surfactant Protein D (SP-D) is designed for use in quantitative ELISA applications.
- Species Reactivity: Human
- Detection Method: Colorimetric - 450nm (TMB)
- Storage Conditions: Store unopened product at 2°C to 8°C
- Intended Use: For research use only
Specifications
Specifications
| Product Type | ELISA Control |
| Quantity | 1 Ea. |
| Species | Human |
| Binding Target | SP-D |
| Gene ID (Entrez) | 6441 |
| Product Line | R&D Systems |
| Synonym | COLEC7collectin-7, collectin 7, Collectin-7, Lung surfactant protein D, PSPD, PSP-D, SFTP4, SFTP4pulmonary surfactant-associated protein D, SFTPD, SPD, SP-Dpulmonary surfactant apoprotein, surfactant protein D, surfactant, pulmonary-associated protein D, surfactant-associated protein, pulmonary 4 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Research Area | Immunology |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
