Learn More
Description
The QuicKit Immunoassay Control Set 1198 is designed for the quantitative measurement of human FAP (also known as seprase) by ELISA.
- Analyte: Human FAP
- Detection Method: Colorimetric (450 nm TMB)
- Format: Lyophilized
- Reconstitution Volume: 2.0 mL
- Contents: 1 vial (low) and 1 vial (high)
- Storage: Store unopened at 2°C to 8°C
Specifications
Specifications
| Product Type | ELISA Control |
| Quantity | 1 Each |
| Species | Human |
| Binding Target | Fibroblast Activation Protein alpha/FAP |
| Gene ID (Entrez) | 2191 |
| Product Line | R&D Systems |
| Synonym | 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAP, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Research Area | Oncology |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
