Learn More
Hardy Diagnostics™ QuickSlide™ Replacement Tubing Kit for GramPRO 80 Automated Gram Stainer
Shop All Hardy Diagnostics Products
Click to view available options
Product Type:
Replacement Tubing Kit
Description
- Requires tube replacement every 1100 cycles
- Icludes batch number and a serial number that is entered under the consumable menu of the unit
- Comes with complete instructions and installation can be done in just a few minutes
- Easy and convenient
Specifications
Specifications
Product Type | Replacement Tubing Kit |
Description | Weight: 1 lb.Dimensions: L X W X H: 30.4 x 17.7 x 7.62cm (12 X 7 X 3 in.) |
For Use With (Equipment) | MGS-80, MGS80-RC-9000, MGS80-SC-8000, MGS80-SK-6000 |