Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel AmpliVue™ Bordetella Assay Kits
SDP

Catalog No. p-4950296 Shop All QuidelOrtho Products
Click to view available options
Type:
Control
Starter Kit
This item is not returnable. View return policy
This item is not returnable. View return policy

In vitro diagnostic test for the qualitative detection of Bordetella pertussis nucleic acids

  • Small, self-contained hand-held cassette
  • Convenient, easy-to-use, disposable device
  • HDA technology; Rapid method of nucleic acid amplification that does not require thermal cycler
  • Lyophilized reagents; Easy-to-use format, just rehydrate and add sample
  • Reagents are stable at higher temperatures than liquid reagents; Small easy-to-use heat blocks
  • Eliminates need for dedicated molecular space and costly capital equipment; Allows for testing in smaller labs
  • CLIA moderately complex
  • Requires minimal training
  • Refrigerated storage; No freezer needed; 2°C to 8°C storage
  • Long shelf life; 2 years from date of manufacture
  • Room temperature set up; No ice or cooling block required

Specifications

Content And Storage Controls storage conditions: 2°C to 8°C
Sample Type Nasopharyngeal Swab
Type Control
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.