Learn More
Description
Includes
Materials for 25 tests with external positive and negative controls
Provides accurate and easy-to-interpret results in 12 minutes or less
- CLIA Complexity: Non-Waived
- CPT Code: 87810
- Sensitivity: 92% (Overall)
- Specificity: 99% (Overall)
- Lateral-flow immunoassay for the qualitative detection of Chlamydia trachomatis antigen directly from endocervical swab or cytology brush specimens
- Simple extraction procedure requires the addition of only two reagents
- Two-color endpoint for easy-to-read results
- Highly accurate in asymptomatic patient population
- Built-in controls confirm test has been performed correctly
- For in vitro diagnostic use
Specifications
Specifications
| Content And Storage | Room Temperature |
| Clia Complexity | Non-waived |
| Detectable Analytes | Chlamydia trachomatis Antigen |
| Sensitivity | 92% |
| Type | Chlamydia Testing Kit |
| Includes | Materials for 25 Tests with external positive and negative control |
| Quantity | 25 Tests |
| Specificity | 0.99 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
