Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Quidel QuickVue™ Chlamydia Test

Click to view available options
Type:
Chlamydia Swab
Chlamydia Testing Kit
Description
Includes
Materials for 25 tests with external positive and negative controls
Provides accurate and easy-to-interpret results in 12 minutes or less
- CLIA Complexity: Non-Waived
- CPT Code: 87810
- Sensitivity: 92% (Overall)
- Specificity: 99% (Overall)
- Lateral-flow immunoassay for the qualitative detection of Chlamydia trachomatis antigen directly from endocervical swab or cytology brush specimens
- Simple extraction procedure requires the addition of only two reagents
- Two-color endpoint for easy-to-read results
- Highly accurate in asymptomatic patient population
- Built-in controls confirm test has been performed correctly
- For in vitro diagnostic use
Specifications
Specifications
Content And Storage | Room Temperature |
Type | Chlamydia Swab |
Quantity | 25 Tests |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction